Tribenoside impurity D
Synonym(s):
Benzyl ether, Dibenzyl ether
Linear Formula:
(C6H5CH2)2O
CAS Number:
Molecular Weight:
198.26
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing