
trans-3-Octen-2-one

Linear Formula:
CH3(CH2)3CH=CHCOCH3
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing