
Sodium barbiturate

Synonym(s):
Barbituric acid sodium salt, Sodium barbiturate
Empirical Formula (Hill Notation):
C4H3N2NaO3
CAS Number:
Molecular Weight:
150.07
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing