
Rhynchophylline

Synonym(s):
Mitrinermine
Empirical Formula (Hill Notation):
C22H28N2O4
CAS Number:
Molecular Weight:
384.47
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing