Skip to Content
Merck
PRT4165 ≥98% (HPLC)

PRT4165

PRT4165 ≥98% (HPLC)
Synonym(s):
2-(3-Pyridinylmethylene)-1H-indene-1,3(2H)-dione; 2-(3-Pyridylmethylene)-1,3-indandione, NSC 600157
Empirical Formula (Hill Notation):
C15H9NO2
CAS Number:
Molecular Weight:
235.24

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing