Propylthiouracil impurity A
Synonym(s):
Thiourea, Sulfourea, Thiocarbamide
Linear Formula:
NH2CSNH2
CAS Number:
Molecular Weight:
76.12
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing