Skip to Content
Merck
Isoamyl acetate ≥95%, FCC, FG

Isoamyl acetate

Isoamyl acetate ≥95%, FCC, FG
Synonym(s):
Isopentyl acetate, Acetic acid 3-methylbutyl ester, Isoamyl acetate
Linear Formula:
CH3COOCH2CH2CH(CH3)2
CAS Number:
Molecular Weight:
130.18
EC Number:

Products

Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing