Insulin (bovine)
Synonym(s):
Insulin from bovine pancreas
Empirical Formula (Hill Notation):
C254H377N65O75S6
CAS Number:
Molecular Weight:
5733.49
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing