![Hydroxyethyl salicylate European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/171/070/d8b7968c-368e-47fc-9f8a-8d0454afb8d7/640/d8b7968c-368e-47fc-9f8a-8d0454afb8d7.png)
Hydroxyethyl salicylate
![Hydroxyethyl salicylate European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/171/070/d8b7968c-368e-47fc-9f8a-8d0454afb8d7/640/d8b7968c-368e-47fc-9f8a-8d0454afb8d7.png)
Synonym(s):
Ethylene glycol monosalicylate, Ethylene glycol salicylate, Salicylic acid 2-hydroxyethyl ester, Salicylic acid ethylene glycol ester
Linear Formula:
HOC6H4COOCH2CH2OH
CAS Number:
Molecular Weight:
182.17
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing