
Fosthiazate

Synonym(s):
O-Ethyl S-(1-methylpropyl) 2-oxo-3-thiazolidinylphosphonothioate, S-sec-Butyl O-ethyl 2-oxo-3-thiazolidinylphosphonothioate
Empirical Formula (Hill Notation):
C9H18NO3PS2
CAS Number:
Molecular Weight:
283.35
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2