
Eupalitin 3-galactoside

Synonym(s):
Eupalitin 3-O-β-D-galactopyranoside, D-Galactopyranosyloxy)-5-hydroxy-2-(4-hydroxyphenyl)-6,7-dimethoxy-4H-1-benzopyran-4-one, Betuletrin, Eupalitin-3-O-galactoside
Empirical Formula (Hill Notation):
C23H24O12
CAS Number:
Molecular Weight:
492.43
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing