
Etodolac impurity mixture

Synonym(s):
Etodolac, 1,8-Diethyl-1,3,4,9-tetrahydropyrano[3,4-b]indole-1-acetic acid, Etodolic acid, NSC 282126
Empirical Formula (Hill Notation):
C17H21NO3
CAS Number:
Molecular Weight:
287.35
Products
Brand
Formula Weight
Manufacturer
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing