![Endoglycosidase F1 from Elizabethkingia miricola recombinant, expressed in E. coli, ≥16 U/mg, buffered aqueous solution](/deepweb/assets/sigmaaldrich/product/images/218/002/ae3f4960-6cce-40cd-8dd7-a43af6cfeb9f/640/ae3f4960-6cce-40cd-8dd7-a43af6cfeb9f.jpg)
Endoglycosidase F1 from Elizabethkingia miricola
![Endoglycosidase F1 from Elizabethkingia miricola recombinant, expressed in E. coli, ≥16 U/mg, buffered aqueous solution](/deepweb/assets/sigmaaldrich/product/images/218/002/ae3f4960-6cce-40cd-8dd7-a43af6cfeb9f/640/ae3f4960-6cce-40cd-8dd7-a43af6cfeb9f.jpg)
Synonym(s):
Endo F1, Endo-β-N-acetylglucosaminidase F1, Endoglycosidase F1 from Chryseobacterium meningosepticum, Endoglycosidase F1 from Elizabethkingia meningoseptica, Endoglycosidase F1 from Flavobacterium meningosepticum
Products
Product Category
Brand
Physical Form
Recombinant Host
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing