(+)-Dihydrocinchonine
Synonym(s):
Hydrocinchonine, Dihydrocinchonine
Empirical Formula (Hill Notation):
C19H24N2O
CAS Number:
Molecular Weight:
296.41
Products
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing