5-bromobenzothiazole
Empirical Formula (Hill Notation):
C7H4BrNS
CAS Number:
Molecular Weight:
214.08
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing