Skip to Content
Merck
2,3-Butanedione analytical standard

2,3-Butanedione

2,3-Butanedione analytical standard
Synonym(s):
Biacetyl, Diacetyl
Linear Formula:
CH3COCOCH3
CAS Number:
Molecular Weight:
86.09
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Gene Alias
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing