Skip to Content
Merck
All Photos(1)

Key Documents

I6893

Sigma-Aldrich

Indoxyl β-D-glucoside

BioXtra, ≥97%

Synonym(s):

3-(β-D-Glucosido)indole, Plant indican

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H17NO6
CAS Number:
Molecular Weight:
295.29
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.55

product line

BioXtra

Assay

≥97%

form

powder

impurities

≤0.0005% Phosphorus (P)

ign. residue

<0.1%

solubility

H2O: 0.1 M, clear to slightly hazy

anion traces

chloride (Cl-): ≤0.05%
sulfate (SO42-): ≤0.05%

cation traces

Al: ≤0.005%
Ca: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.0005%
NH4+: ≤0.05%
Na: ≤0.05%
Pb: ≤0.001%
Zn: ≤0.0005%

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@@H](Oc2c[nH]c3ccccc23)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C14H17NO6/c16-6-10-11(17)12(18)13(19)14(21-10)20-9-5-15-8-4-2-1-3-7(8)9/h1-5,10-19H,6H2/t10-,11-,12+,13-,14-/m1/s1

InChI key

XVARCVCWNFACQC-RKQHYHRCSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Bradd J Haley et al.
Applied and environmental microbiology, 75(5), 1248-1255 (2009-01-07)
Salmonella outbreaks from contaminated water and nonanimal foods (e.g., produce) are increasingly reported. To address the environment as a potential source of pathogenic Salmonella, we investigated levels of salmonellae and the geographic and temporal variation of Salmonella serotypes from surface
Thierry Maugard et al.
Biotechnology progress, 18(5), 1104-1108 (2002-10-05)
In this article, a HPLC method to identify and quantify the dyes and the indigo precursors produced in Polygonum tinctorium is described. Using this technique, indican has been positively identified in extracts of P. tinctorium. Our work with two cultivars
[Determination of indican in Polygonum tinctorium ait. in various growth periods].
H J Wang et al.
Zhong yao tong bao (Beijing, China : 1981), 13(11), 11-13 (1988-11-01)
R Armon et al.
Canadian journal of microbiology, 34(1), 78-79 (1988-01-01)
Medium m-CP, designed for the isolation of Clostridium perfringens from water samples, contains indoxyl beta-D-glucoside, an expensive chemical that is present at a high concentration in this medium. The use of m-CP with three concentrations of indoxyl beta-D-glucoside was tested
A N Ley et al.
Canadian journal of microbiology, 34(5), 690-693 (1988-05-01)
About 97% of Escherichia coli strains produce beta-glucuronidase, but almost all other Enterobacteriaceae lack this enzyme. A D-glucopyranosiduronic acid (glucuronide) possessing a readily detectable beta-linked aglycone should, therefore, constitute a specific reagent for the detection of this organism. For this

Articles

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service