Skip to Content
Merck
All Photos(1)

Key Documents

COMH421174ED

Combi-Blocks, Inc.

Boc-l-thr(bzl)-o-ch2-f-ch2-cooh dcha

Sign Into View Organizational & Contract Pricing


About This Item

Molecular Weight:
638.84
MDL number:

Iupac Name

dicyclohexylamine 2-(4-(((O-benzyl-N-(tert-butoxycarbonyl)-L-threonyl)oxy)methyl)phenyl)acetate

Inchi Code

1S/C25H31NO7.C12H23N/c1-17(31-15-19-8-6-5-7-9-19)22(26-24(30)33-25(2,3)4)23(29)32-16-20-12-10-18(11-13-20)14-21(27)28;1-3-7-11(8-4-1)13-12-9-5-2-6-10-12/h5-13,17,22H,14-16H2,1-4H3,(H,26,30)(H,27,28);11-13H,1-10H2/t17-,22+;/m1./s1

InChI key

BSLDLXPANSHTOC-YITVUBHWSA-N

Purity

98%

Country of Origin

CN

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?HA-4913

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?HA-4913


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service