Skip to Content
Merck
All Photos(1)

Key Documents

CIAH98B928A1

ChemScene LLC (Preferred Partner)

3-(4-Methylnaphthalen-1-yl)propanoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
874,15 kr
1 G
1 833,35 kr

Vendor SKU: CS-0037904-250MG

874,15 kr


Ships in 14 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
250 MG
874,15 kr
1 G
1 833,35 kr

About This Item

CAS Number:
Molecular Weight:
214.26
MDL number:

Vendor SKU: CS-0037904-250MG

874,15 kr


Ships in 14 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.

bp

215-220 C at 5 mmHg

mp

158 C

Iupac Name

3-(4-methyl-1-naphthyl)propanoic acid

Inchi Code

1S/C14H14O2/c1-10-6-7-11(8-9-14(15)16)13-5-3-2-4-12(10)13/h2-7H,8-9H2,1H3,(H,15,16)

InChI key

NLJSIZYZZYSWLL-UHFFFAOYSA-N

storage temp.

4C

Purity

98%

Country of Origin

CN

Physical Form

Solid

Product Link

https://www.chemscene.com/76673-34-2.html

Boiling point

215-220 C at 5 mmHg

Melting Point

158 C

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://file.chemscene.com/pdf/UsaMSDS/MSDSUSACS-0037904.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service