Merck
  • HPLC Analysis to Determine the Half-life and Bioavailability of the Termiticides Bifenthrin and Fipronil in Soil.
HPLC Analysis to Determine the Half-life and Bioavailability of the Termiticides Bifenthrin and Fipronil in Soil.
Journal of economic entomology (2017-10-19)
F Manzoor, M Pervez
ABSTRACT
The aim of this study was to test the bioavailability and degradation in soil of the termiticides bifenthrin and fipronil, which are used to treat subterranean termites (Heterotermes indicola, Wasmann). Soil collected from different areas of Lahore was categorized as sandy clay loam (SCL) or sandy loam (SL). Laboratory bioassays were conducted to determine the bioavailability ratio of bifenthrin and fipronil in each type of soil after different periods of time. LT50 values were determined posttreatment at different time intervals. Regarding soil type, both termiticides were more effective in SL soil, compared with SCL soil posttreatment. There were significant differences in termite mortality in treated compared with untreated control samples (P < 0.005). To test the degradation rate of soil termiticides, both qualitative and quantitative analyses were carried out by HPLC, and the effect of time on termiticide recovery rate determined. Calculated half-life values for bifenthrin (maximum, 1,002 and 1,262 d in SCL soil and SL soil, respectively) indicated that it persisted in both soil types at all concentrations. The maximum calculated half-life values of fipronil were 270 and 555 d in SCL and SL soil, respectively. At lower concentrations and over longer periods of time, fipronil completely degraded in SL soil, while a negligible amount was detected in SCL soil. Termiticide concentration decreased over time, as did the termiticide recovery rate. Overall, bifenthrin was more persistent than fipronil under all treatment conditions tested.
Sigma-Aldrich
3-Phenyl-1H-pyrazole, 97%