Merck
  • Medical complications during inpatient rehabilitation among patients with traumatic disorders of consciousness.
Medical complications during inpatient rehabilitation among patients with traumatic disorders of consciousness.
Archives of physical medicine and rehabilitation (2013-06-06)
John Whyte, Annette M Nordenbo, Kathleen Kalmar, Barbara Merges, Emilia Bagiella, Helena Chang, Stuart Yablon, Sooja Cho, Flora Hammond, Allen Khademi, Joseph Giacino
ABSTRACT
To assess the incidence of medical complications in patients with recent traumatic disorders of consciousness (DOCs). Data on adverse events in a placebo controlled trial of amantadine hydrochloride revealed no group difference, which allowed these events to be reanalyzed descriptively as medical complications experienced by the 2 groups collectively. Eleven clinical facilities in the United States, Denmark, and Germany with specialty rehabilitation programs for patients with DOCs. Patients (N=184) with nonpenetrating traumatic brain injury enrolled from acute inpatient rehabilitation programs between 4 and 16 weeks postinjury. Participants were randomized to receive 200 to 400mg of amantadine hydrochloride or placebo daily for 4 weeks, and followed for an additional 2 weeks. Adverse events were recorded and categorized with respect to their nature, timing, and severity. Number, type, and severity of medical complications occurring during the 6-week study interval. A total of 468 medical complications were documented among the patients (.40 events per week per patient). More than 80% of patients experienced at least 1 medical complication, and 41 of these were defined as serious adverse events. New medical complications declined over time in rehabilitation and were not dependent on time since injury. Hypertonia, agitation/aggression, urinary tract infection, and sleep disturbance were the most commonly reported problems. Hydrocephalus, pneumonia, gastrointestinal problems, and paroxysmal sympathetic hyperactivity were the most likely to be severe. Patients with DOCs have a high rate of medical complications early after injury. Many of these complications require brain injury expertise for optimal management. Active medical management appears to contribute to the reduction in new complications. An optimal system of care for DOC patients must provide expert medical management in the early weeks after injury.
Sigma-Aldrich
Amantadine hydrochloride
Pricing and availability is not currently available.