Merck
  • Defining genome-wide CRISPR-Cas genome-editing nuclease activity with GUIDE-seq.
Defining genome-wide CRISPR-Cas genome-editing nuclease activity with GUIDE-seq.
Nature protocols (2021-11-14)
Nikolay L Malinin, GaHyun Lee, Cicera R Lazzarotto, Yichao Li, Zongli Zheng, Nhu T Nguyen, Matthew Liebers, Ved V Topkar, A John Iafrate, Long P Le, Martin J Aryee, J Keith Joung, Shengdar Q Tsai
ABSTRACT
Genome-wide unbiased identification of double-stranded breaks enabled by sequencing (GUIDE-seq) is a sensitive, unbiased, genome-wide method for defining the activity of genome-editing nucleases in living cells. GUIDE-seq is based on the principle of efficient integration of an end-protected double-stranded oligodeoxynucleotide tag into sites of nuclease-induced DNA double-stranded breaks, followed by amplification of tag-containing genomic DNA molecules and high-throughput sequencing. Here we describe a detailed GUIDE-seq protocol including cell transfection, library preparation, sequencing and bioinformatic analysis. The entire protocol including cell culture can be completed in 9 d. Once tag-integrated genomic DNA is isolated, library preparation, sequencing and analysis can be performed in 3 d. The result is a genome-wide catalog of off-target sites ranked by nuclease activity as measured by GUIDE-seq read counts. GUIDE-seq is one of the most sensitive cell-based methods for defining genome-wide off-target activity and has been broadly adopted for research and therapeutic use.
Sigma-Aldrich
Tetramethylammonium chloride solution, Molecular Biology