Merck
Revealing RCOR2 as a regulatory component of nuclear speckles.
Epigenetics & chromatin (2021-11-26)
Carlos Rivera, Daniel Verbel-Vergara, Duxan Arancibia, Anna Lappala, Marcela González, Fabián Guzmán, Gianluca Merello, Jeannie T Lee, María Estela Andrés
ABSTRACT
Nuclear processes such as transcription and RNA maturation can be impacted by subnuclear compartmentalization in condensates and nuclear bodies. Here, we characterize the nature of nuclear granules formed by REST corepressor 2 (RCOR2), a nuclear protein essential for pluripotency maintenance and central nervous system development. Using biochemical approaches and high-resolution microscopy, we reveal that RCOR2 is localized in nuclear speckles across multiple cell types, including neurons in the brain. RCOR2 forms complexes with nuclear speckle components such as SON, SRSF7, and SRRM2. When cells are exposed to chemical stress, RCOR2 behaves as a core component of the nuclear speckle and is stabilized by RNA. In turn, nuclear speckle morphology appears to depend on RCOR2. Specifically, RCOR2 knockdown results larger nuclear speckles, whereas overexpressing RCOR2 leads to smaller and rounder nuclear speckles. Our study suggests that RCOR2 is a regulatory component of the nuclear speckle bodies, setting this co-repressor protein as a factor that controls nuclear speckles behavior.
Sigma-Aldrich
Anti-Splicing Factor SC-35 antibody, Mouse monoclonal, clone SC-35, purified from hybridoma cell culture
Sigma-Aldrich
Anti-ASF-1 Antibody, clone AK-96, clone AK-96, from mouse
Sigma-Aldrich
Anti-RCOR2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution