Merck
  • Facile synthesis of silver nanoparticles useful for fabrication of high-conductivity elements for printed electronics.
Facile synthesis of silver nanoparticles useful for fabrication of high-conductivity elements for printed electronics.
Journal of the American Chemical Society (2005-03-10)
Yuning Li, Yiliang Wu, Beng S Ong
ABSTRACT
A facile synthesis of stable silver nanoparticles having a particle size of <10 nm is described. The synthesis involved reduction of silver acetate with a substituted hydrazine, such as PhNHNH2, in the presence of a 1-alkylamine, such as C16H33NH2, in toluene at 25-60 degrees C. Spin-coated thin films or printed electronic features of alkylamine-stabilized silver nanoparticles could be easily converted at 120-160 degrees C into highly conductive films or elements with conductivity of 2-4 x 104 S cm-1. Organic thin-film transistors with printed silver source/drain electrodes of this nature exhibited field-effect transistor properties which are similar to those of the devices using vacuum-deposited silver electrodes.
Sigma-Aldrich
Silver (I) Acetate, anhydrous, ≥99.9% trace metals basis