Merck
Angiogenesis inhibitors derived from thalidomide.
Bioorganic & medicinal chemistry letters (2005-09-27)
Tomomi Noguchi, Haruka Fujimoto, Hiroko Sano, Atsushi Miyajima, Hiroyuki Miyachi, Yuichi Hashimoto
ABSTRACT
5-Hydroxy-2-(2,6-diisopropylphenyl)-1H-isoindole-1,3-dione (5HPP-33: 10), which was obtained during our previous structural development studies on thalidomide, was revealed to possess potent anti-angiogenic activity in a human umbilical vein endothelial cell (HUVEC) assay. Thalidomide (1) and its metabolite, 5-hydroxythalidomide (5-HT: 2), which possesses a hydroxyl group at the position corresponding to that of 5HPP-33, as well as IMiDs (immunomodulatory derivatives of thalidomide: 3 and 5), also showed weak or moderate activity in the same assay.
Sigma-Aldrich
(±)-Thalidomide, ≥98%, powder