Merck
  • Enabling lead discovery for histone lysine demethylases by high-throughput RapidFire mass spectrometry.
Enabling lead discovery for histone lysine demethylases by high-throughput RapidFire mass spectrometry.
Journal of biomolecular screening (2011-08-24)
Sue E Hutchinson, Melanie V Leveridge, Michelle L Heathcote, Peter Francis, Laura Williams, Michelle Gee, Jordi Munoz-Muriedas, Bill Leavens, Anthony Shillings, Emma Jones, Paul Homes, Stuart Baddeley, Chun-Wa Chung, Angela Bridges, Argyrides Argyrou
ABSTRACT
A high-throughput RapidFire mass spectrometry assay is described for the JMJD2 family of Fe(2+), O(2), and α-ketoglutarate-dependent histone lysine demethylases. The assay employs a short amino acid peptide substrate, corresponding to the first 15 amino acid residues of histone H3, but mutated at two positions to increase assay sensitivity. The assay monitors the direct formation of the dimethylated-Lys9 product from the trimethylated-Lys9 peptide substrate. Monitoring the formation of the monomethylated and des-methylated peptide products is also possible. The assay was validated using known inhibitors of the histone lysine demethylases, including 2,4-pyridinedicarboxylic acid and an α-ketoglutarate analogue. With a sampling rate of 7 s per well, the RapidFire technology permitted the single-concentration screening of 101 226 compounds against JMJD2C in 10 days using two instruments, typically giving Z' values of 0.75 to 0.85. Several compounds were identified of the 8-hydroxyquinoline chemotype, a known series of inhibitors of the Lys9-specific histone demethylases. The peptide also functions as a substrate for JMJD2A, JMJD2D, and JMJD2E, thus enabling the development of assays for all 3 enzymes to monitor progress in compound selectivity. The assay represents the first report of a RapidFire mass spectrometry assay for an epigenetics target.
Sigma-Aldrich
2,4-Pyridinedicarboxylic acid, ≥98.0%
Millipore
Protease Inhibitor Cocktail Set III, EDTA-Free, Protease inhibitor cocktail III, EDTA-free for inhibiting aspartic, cysteine, and serine proteases as well as aminopeptidases in mammalian cells and tissues.