Merck
A fucose-containing O-glycoepitope on bovine and human nucleolin.
Glycobiology (2008-11-22)
Silvia Aldi, Cinzia Della Giovampaola, Riccardo Focarelli, Alessandro Armini, Marina Ziche, Federica Finetti, Floriana Rosati
ABSTRACT
In this paper, we demonstrate the existence and localization of fucosyl-containing O-glycoforms of nucleolin in cultured bovine endothelial cells (CVEC) and malignant cultured human A431 cells. The tool for this discovery was an antibody raised against gp273, a glycoprotein ligand for the sperm-egg interaction in the mollusc bivalve Unio elongatulus. The function and immunological properties of gp273 mainly depend on clustered Lewis-like, fucose-containing O-glycans. Here an anti-gp273 antibody was used to evaluate whether glycoepitopes similar to those of gp273 are part of potential ligands of selectins in endothelial cells. We found that anti-gp273 strongly and exclusively interacted with a 110 kDa protein in CVEC and A431 tumor cells. After partial purification, mass spectrometry identified the protein as nucleolin. This was confirmed by comparing anti-gp273 and anti-nucleolin antibody immunoblotting after nucleolin depletion. We confirmed that anti-gp273 binding to nuclear and extranuclear nucleolin was against a fucose-containing O-glycoepitope by immunoblot analysis of the protein after chemically removing O-glycans and by lectin-blot analysis of control and nucleolin-depleted samples. Using anti-gp273 IgG, we detected nucleolin on the plasma membrane and cytoplasm. O-Glycosylation may regulate the plethora of functions in which nucleolin is involved.