
Secologanin

Empirical Formula (Hill Notation):
C17H24O10
CAS Number:
Molecular Weight:
388.37
Products
Brand
Formula Weight
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing