
Oxybuprocaine hydrochloride

Synonym(s):
Benoxinate hydrochloride, 2-(Diethylamino)ethyl 4-amino-3-butoxybenzoate hydrochloride, 4-Amino-3-butoxybenzoic acid diethylaminoethyl ester, Oxybuprocaine hydrochloride
Empirical Formula (Hill Notation):
C17H28N2O3 · HCl
CAS Number:
Molecular Weight:
344.88
Products
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2