
Malvidin 3,5-diglucoside chloride

Synonym(s):
Malvin(chloride), Malvidin chloride 3,5-diglucoside, Malvidin-3,5-diglucoside, Malvoside
Empirical Formula (Hill Notation):
C29H35ClO17
Products
Formula Weight
Manufacturer
Product Line
Purity
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing