
Dichlorprop-methyl ester

Synonym(s):
Methyl 2-(2,4-dichlorophenoxy)propionate
Empirical Formula (Hill Notation):
C10H10Cl2O3
CAS Number:
Molecular Weight:
249.09
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing