HPLC Analysis of Mianserin Enantiomers on Astec® CHIROBIOTIC® V2
Materials
analytical column
Product No.
Description
Pricing
Application
Product No.
Description
Pricing
standard
Product No.
Description
Pricing
CONDITIONS
column
CHIROBIOTIC V2, 25 cm x 4.6 mm I.D., 5 μm particles (15024AST)
mobile phase
15 mM ammonium acetate in methanol
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 230 nm
injection
5 μL
sample
1 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC