Merck
  • Berberine Depresses Contraction of Smooth Muscle via Inhibiting Myosin Light-chain Kinase.
Berberine Depresses Contraction of Smooth Muscle via Inhibiting Myosin Light-chain Kinase.
Pharmacognosy magazine (2017-08-26)
Zhili Xu, Mingbo Zhang, Deqiang Dou, Xiaojun Tao, Tingguo Kang
ABSTRACT
Berberine is a natural isoquinoline alkaloid possessing various pharmacological effects, particularly apparent in the treatment of diarrhea, but the underlying mechanism remains unclear. Smooth muscle myosin light-chain kinase (MLCK) plays a crucial role in the smooth muscle relaxation-contraction events, and it is well known that berberine can effectively depress the contraction of smooth muscle. Hence, whether berberine could inhibit MLCK and then depress the smooth muscle contractility might be researched. The purpose of this study is to investigate the effects of berberine on MLCK. Based on this, the contractility of gastro-intestine, catalysis activity of MLCK, and molecular docking are going to be evaluated. The experiment of smooth muscle contraction was directly monitored the contractions of the isolated gastrointestine by frequency and amplitude at different concentration of berberine. The effects of berberine on MLCK were measured in the presence of Ca The phosphorylation of myosin and the Mg The present work gives a deep insight into the molecular mechanism for the treatment of diarrhea with berberine, i.e., berberine could suppress the contractility of smooth muscle through binding to MLCK and depressing the catalysis activity of MLCK. Berberine significantly reduced the amplitude of contraction in isolated duodenum and gastric strips in ratsBerberine inhibited the phosphorylated extents of MLC20 and Mg2+-ATPase activity of phosphorylated myosin induced by MLCKBerberine binds to the ATP binding site of MLCK by hydrophobic effect and hydrogen bondingBerberine may modulate contraction of smooth muscle by inhibiting MLCK.