Merck
  • Social context has differential effects on acquisition of nicotine self-administration in male and female rats.
Social context has differential effects on acquisition of nicotine self-administration in male and female rats.
Psychopharmacology (2017-04-01)
Natalie A Peartree, Kayla N Hatch, Julianna G Goenaga, Nora R Dado, Hanna Molla, Martin A Dufwenberg, Allegra Campagna, Rachel Mendoza, Timothy H C Cheung, Joshua S Talboom, Janet L Neisewander
ABSTRACT
Smoking typically begins during adolescence or early adulthood in a social context, yet the role of social context in animal models is poorly understood. The present study examined the effect of social context on acquisition of nicotine self-administration. Sixty-day-old male and female Sprague-Dawley rats were trained to press a lever for nicotine (0.015 mg/kg, IV) or saline infusions (males only) on a fixed ratio (FR1) schedule of reinforcement across nine sessions in duplex chambers that were conjoined with either a solid wall or a wall containing wire mesh creating a social context between rat dyads (social visual, auditory, and olfactory cues). In a subsequent experiment, sex differences and dose-dependent effects of nicotine [0 (saline), 0.015 or 0.03 mg/kg, IV] were directly compared in rats trained in the isolated or social context on a schedule progressing from FR1 to FR3. These rats were given 20 sessions followed by 3 extinction sessions. We consistently found transient social facilitation of low-dose nicotine self-administration in males during the first session. However, across training overall, we found social suppression of nicotine intake that was most prominent in females during later sessions. Collectively, these findings suggest that at the age of transition from adolescence to adulthood, a social context enhances the initial reinforcing effects of nicotine in males, but protects against nicotine intake during later sessions especially in females. These findings highlight the importance of sex and social context in studying neural mechanisms involved in initiation of nicotine use.
Sigma-Aldrich
Ticarcillin disodium salt
Pricing and availability is not currently available.