Merck
  • Determination of furanochromones and pyranocoumarins in drugs and Ammi visnaga fruits by combined solid-phase extraction-high-performance liquid chromatography and thin-layer chromatography-high-performance liquid chromatography.
Determination of furanochromones and pyranocoumarins in drugs and Ammi visnaga fruits by combined solid-phase extraction-high-performance liquid chromatography and thin-layer chromatography-high-performance liquid chromatography.
Journal of chromatography. A (1998-05-16)
G Zgórka, T Dragan, K Głowniak, E Basiura
ABSTRACT
A new, simple and rapid solid-phase extraction method for the determination of furanochromones and pyranocoumarins in Ammi visnaga L. fruits and pharmaceuticals by reversed-phase high-performance liquid chromatography (RP-HPLC) was developed. The isolation of compounds examined was carried out on octadecyl BakerBond SPE columns using various concentrations of methanol, acetonitrile and tetrahydrofuran in water. High and reproducible recoveries were obtained. To compare the results of quantitative analysis a preparative TLC procedure was also elaborated and carried out.