Merck
Solvent inclusion in form II carbamazepine.
Chemical communications (Cambridge, England) (2007-05-29)
Aurora J Cruz Cabeza, Graeme M Day, W D Samuel Motherwell, William Jones
ABSTRACT
We report on experimental and theoretical evidence for solvent inclusion in form II carbamazepine (R3) and discuss the implications for the formation and stability of this form.
Sigma-Aldrich
Tridecane, ≥99%
Pricing and availability is not currently available.
Supelco
Tridecane, analytical standard
Pricing and availability is not currently available.