Merck
  • Preparation and characterization of pH-responsive polymeric micelles for the delivery of photosensitizing anticancer drugs.
Preparation and characterization of pH-responsive polymeric micelles for the delivery of photosensitizing anticancer drugs.
Journal of pharmaceutical sciences (2000-02-09)
J Taillefer, M C Jones, N Brasseur, J E van Lier, J C Leroux
ABSTRACT
pH-responsive polymeric micelles (PM) consisting of random copolymers of N-isopropylacrylamide (NIPA), methacrylic acid (MAA), and octadecyl acrylate (ODA) were prepared and characterized. The critical aggregation concentration, as determined by a fluorescence probe technique, was approximately 10 mg/L in water and phosphate-buffered saline. Phase transition pH was estimated at 5.7. The decrease in pH was accompanied by the destruction of hydrophobic clusters. Micelle size was dependent on temperature and the nature of the aqueous medium. The micelles were successfully loaded with a substantial amount of a photoactive anticancer drug, namely, aluminum chloride phthalocyanine (AlClPc). pH-responsive PM loaded with AlClPc were found to exhibit higher cytotoxicity against EMT-6 mouse mammary cells in vitro than control Cremophor EL formulation. These results show the potential of poly(NIPA-co-MAA-co-ODA) for in vivo administration of water-insoluble, photosensitizing anticancer drugs.