Merck
  • Synthesis and characterization of new potent TLR7 antagonists based on analysis of the binding mode using biomolecular simulations.
Synthesis and characterization of new potent TLR7 antagonists based on analysis of the binding mode using biomolecular simulations.
European journal of medicinal chemistry (2020-11-16)
Sourav Pal, Barnali Paul, Purbita Bandopadhyay, Nagothy Preethy, Dipika Sarkar, Oindrila Rahaman, Sunny Goon, Swarnali Roy, Dipyaman Ganguly, Arindam Talukdar
ABSTRACT
Aberrant activation of the endosomal Toll-like receptor 7 (TLR7) has been implicated in myriad autoimmune diseases and is an established therapeutic target in such conditions. Development of diverse TLR7 antagonists is mainly accomplished through random screening. To correlate human TLR7 (hTLR7) antagonistic activity with the structural features in different chemotypes, we derived a hypothetical binding model based on molecular docking analysis along with molecular dynamics (MD) simulations study. The binding hypothesis revealed different pockets, grooves and a central cavity where ligand-receptor interaction with specific residues through hydrophobic and hydrogen bond interactions take place, which correlate with TLR7 antagonistic activity thus paving the way for rational design using varied chemotypes. Based on the structural insight thus gained, TLR7 antagonists with quinazoline were designed to understand the effect of engagement of these pockets as well as boundaries of the chemical space associated with them. The newly synthesized most potent hTLR7 antagonist, i.e. compound 63, showed IC50 value of 1.03 ± 0.05 μM and was validated by performing primary assay in human plasmacytoid dendritic cells (pDC) (IC50pDC: 1.42 μM). The biological validation of the synthesized molecules was performed in TLR7-reporter HEK293 cells as well as in human plasmacytoid dendritic cells (pDCs). Our study provides a rational design approach thus facilitating further development of novel small molecule hTLR7 antagonists based on different chemical scaffolds.
Sigma-Aldrich
CL264, ≥98% (HPLC)