Merck
  • Newly discovered anti-inflammatory properties of the benzamides and nicotinamides.
Newly discovered anti-inflammatory properties of the benzamides and nicotinamides.
Molecular and cellular biochemistry (1999-05-20)
R W Pero, B Axelsson, D Siemann, D Chaplin, G Dougherty
ABSTRACT
Our laboratory has concentrated on the possible regulation the benzamides and nicotinamides may have on the processes of DNA repair and apoptosis. Recent reports have suggested that both apoptosis and inflammation are regulated by the transcription factor NF-kappaB. We have initiated studies regarding the hypothesis that the benzamides and nicotinamides could inhibit the production of tumor necrosis factor alpha (TNFalpha) and the inflammatory response as well as induce apoptosis via inhibition of NF-kappaB. Our data have shown that nicotinamide and two N-substituted benzamides, metoclopramide (MCA) and 3-chloroprocainamide (3-CPA), gave dose dependent inhibition of lipopolysacharide induced TNFalpha in the mouse within the dose range of 10-500 mg/kg. Moreover, lung edema was prevented in the rat by 3 x 50 mg/kg doses of 3-CPA or MCA, and 100-200 microM doses of MCA could also inhibit NF-kappaB in Hela cells. Taken together these data strongly support the notion that benzamides and nicotinamides have potent anti-inflammatory and antitumor properties, because their primary mechanism of action is regulated by inhibition at the gene transcription level of NF-kappaB, which in turn inhibits TNFalpha and induces apoptosis.
Sigma-Aldrich
Metoclopramide hydrochloride, solid
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, tablet, 20 mg substrate per tablet