Merck
  • Fast and High Strength Soft Tissue Bioadhesives Based on a Peptide Dendrimer with Antimicrobial Properties and Hemostatic Ability.
Fast and High Strength Soft Tissue Bioadhesives Based on a Peptide Dendrimer with Antimicrobial Properties and Hemostatic Ability.
ACS applied materials & interfaces (2019-12-21)
Haofang Zhu, Xingheng Mei, Yiyan He, Hongli Mao, Wenbo Tang, Rong Liu, Jun Yang, Kui Luo, Zhongwei Gu, Lian Zhou
ABSTRACT
Due to the inconvenience of application, risk of extra damage to fragile soft tissues, and the high incidence of late-stage complications, significant research endeavors have been focused on developing safe and effective bioadhesives to replace or assist the traditional suture techniques for wound closure. Here, we describe a fast and high strength bioadhesive based on polysaccharides and peptide dendrimers (OCMC/G3KP) with inherent hemostatic ability and antibacterial properties. Compared with the commercial bioadhesive Coseal, the OCMC/G3KP hydrogel shows a remarkable 5-fold increase in adhesion strength. The in vivo studies further confirm the superior wound healing performance of the OCMC/G3KP hydrogel in contrast with Coseal and conventional sutures. The OCMC/G3KP hydrogels are efficient and biocompatible bioadhesives with precise controllability that could be flexibly modulated to meet diverse clinical demands.
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide, ≥97.0% (T)