Merck
  • Rev-erbα Inhibits Proliferation and Promotes Apoptosis of Preadipocytes through the Agonist GSK4112.
Rev-erbα Inhibits Proliferation and Promotes Apoptosis of Preadipocytes through the Agonist GSK4112.
International journal of molecular sciences (2019-09-25)
Guiyan Chu, Xiaoge Zhou, Yamei Hu, Shengjie Shi, Gongshe Yang
ABSTRACT
Proliferation and apoptosis are important physiological processes of preadipocytes. Rev-erbα is a circadian clock gene, and its activity contributes to several physiological processes in various cells. Previous studies demonstrated that Rev-erbα promotes preadipocyte differentiation, but a role of Rev-erbα on preadipocyte proliferation and apoptosis has not been demonstrated. GSK4112 is often used as an agonist of Rev-erbα. In this study, we used GSK4112 to explore the effects of Rev-erbα on preadipocyte proliferation and apoptosis by RT-qPCR, Western blot, Cell Counting Kit-8 (CCK8) measurement, 5-Ethynyl-2'-deoxyuridine (EdU) staining, Annexin V-FITC/PI staining, and flow cytometry. These results revealed that GSK4112 inhibited the viability of 3T3-L1 preadipocytes and decreased cell numbers. There was also decreased expression of the proliferation-related gene Cyclin D and the canonical Wingless-type (Wnt) signaling effect factor β-catenin. Furthermore, palmitate (PA)-inducing cell apoptosis was promoted. Overall, these results reveal that Rev-erbα plays a role in proliferation and palmitate (PA)-inducing apoptosis of 3T3-L1 preadipocytes, and thus may be a new molecular target in efforts to prevent and treat obesity and related disease.
Sigma-Aldrich
GSK4112, ≥98% (HPLC)
Pricing and availability is not currently available.