Merck
  • Functional nucleoside transporters are required for gemcitabine influx and manifestation of toxicity in cancer cell lines.
Functional nucleoside transporters are required for gemcitabine influx and manifestation of toxicity in cancer cell lines.
Cancer research (1998-10-10)
J R Mackey, R S Mani, M Selner, D Mowles, J D Young, J A Belt, C R Crawford, C E Cass
ABSTRACT
Gemcitabine (2',2'-difluorodeoxycytidine) is a novel pyrimidine nucleoside drug with clinical efficacy in several common epithelial cancers. We have proposed that gemcitabine requires nucleoside transporter (NT) proteins to permeate the plasma membrane and to exhibit pharmacological activity. In humans, there are seven reported distinct NT activities varying in substrate specificity, sodium dependence, and sensitivity to inhibition by nitrobenzylthioinosine (NBMPR) and dipyridamole. To determine which NTs are required for gemcitabine-dependent growth inhibition, cultures from a panel of 12 cell lines with defined plasma membrane NT activities were incubated with different concentrations of gemcitabine. Cell proliferation was assessed by the sulforhodamine B assay and cell enumeration to identify the concentrations of gemcitabine that inhibited cell replication by 50% (IC50s). NT activity was a prerequisite for growth inhibition in vitro because: (a) the nucleoside transport-deficient cells were highly resistant to gemcitabine; and (b) treatment of cells that exhibited only equilibrative NT activity with NBMPR or dipyridamole increased resistance to gemcitabine by 39- to 1800-fold. These data suggested that the type of NT activities possessed by a cell may be an important determinant of its sensitivity to gemcitabine and that NT deficiency may confer significant gemcitabine resistance. We analyzed the uptake kinetics of [3H]gemcitabine by each of five human NT activities in cell lines that exhibited a single NT activity in isolation; transient transfection of the cDNAs encoding the human concentrative NT proteins (hCNT1 and hCNT2) was used to study the cit and cif activities, respectively. The efficiency of gemcitabine uptake varied markedly among the cell lines with single NTs: es approximately = cit > ei > cib > cif. The transportability of [3H]gemcitabine was demonstrated by reconstitution of the human es NT in proteoliposomes, confirming that gemcitabine permeation is a protein-mediated process.