Merck
  • PI3K Inhibition Activates SGK1 via a Feedback Loop to Promote Chromatin-Based Regulation of ER-Dependent Gene Expression.
PI3K Inhibition Activates SGK1 via a Feedback Loop to Promote Chromatin-Based Regulation of ER-Dependent Gene Expression.
Cell reports (2019-04-04)
Eneda Toska, Pau Castel, Sagar Chhangawala, Amaia Arruabarrena-Aristorena, Carmen Chan, Vasilis C Hristidis, Emiliano Cocco, Mirna Sallaku, Guotai Xu, Jane Park, Gerard Minuesa, Sophie G Shifman, Nicholas D Socci, Richard Koche, Christina S Leslie, Maurizio Scaltriti, José Baselga
ABSTRACT
The PI3K pathway integrates extracellular stimuli to phosphorylate effectors such as AKT and serum-and-glucocorticoid-regulated kinase (SGK1). We have previously reported that the PI3K pathway regulates estrogen receptor (ER)-dependent transcription in breast cancer through the phosphorylation of the lysine methyltransferase KMT2D by AKT. Here, we show that PI3Kα inhibition, via a negative-feedback loop, activates SGK1 to promote chromatin-based regulation of ER-dependent transcription. PI3K/AKT inhibitors activate ER, which promotes SGK1 transcription through direct binding to its promoter. Elevated SGK1, in turn, phosphorylates KMT2D, suppressing its function, leading to a loss of methylation of lysine 4 on histone H3 (H3K4) and a repressive chromatin state at ER loci to attenuate ER activity. Thus, SGK1 regulates the chromatin landscape and ER-dependent transcription via the direct phosphorylation of KMT2D. These findings reveal an ER-SGK1-KMT2D signaling circuit aimed to attenuate ER response through a role for SGK1 to program chromatin and ER transcriptional output.
Sigma-Aldrich
Anti-KMT2D antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution