Merck
  • Antimutagenic activity of 5alpha-cholest-7-en-3beta-ol, a new component from the starfish asterina pectinifera.
Antimutagenic activity of 5alpha-cholest-7-en-3beta-ol, a new component from the starfish asterina pectinifera.
Biological & pharmaceutical bulletin (2000-10-21)
Y H Han, J H Ham, N J Lee, C H Park, Y H Shin, D U Lee
ABSTRACT
From the butanol fraction of the starfish Asterina pectinifera Müler et Troschel (Asteriidae), we have isolated a new component, 5alpha-cholest-7-en-3beta-ol. Its antigenotoxic and antimutagenic activities were examined by the SOS chromotest with Escherichia coli PQ37 and by Ames test with Salmonella typhimurium TA1538, respectively. 5alpha-Cholest-7-en-3beta-ol showed potent antigenotoxic activity against the mutagens, both MNNG and NQO. For 100% of antigenotoxicity, the concentration of the compound applied against MNNG and NQO were 10 microg and 5 microg per reaction tube, respectively. Its antimutagenic activity with S. typhimurium TA1538 against the mutagen MNNG was very effective. When its concentrations were varied from 1 up to 10 microg dose per plate, the inhibition ratio of revertant CFU of TA1538 per plate was increased accordingly, from 25.2 to 99.2%. These results suggest that 5alpha-cholest-7-en-3beta-ol possesses antigenotoxic and antimutagenic activity and might be useful as a chemopreventive agent.
Sigma-Aldrich
5α-Cholest-7-en-3β-ol
Pricing and availability is not currently available.