Merck
  • Circular RNAs are differentially expressed in liver ischemia/reperfusion injury model.
Circular RNAs are differentially expressed in liver ischemia/reperfusion injury model.
Journal of cellular biochemistry (2018-05-19)
Zhiqiang Ye, Qinglei Kong, Jianhua Han, Jingyi Deng, Miaolue Wu, Hong Deng
ABSTRACT
Liver ischemia/reperfusion (I/R) injury has high mortality due to the intense inflammatory process occurs in the liver. However, the pathological mechanism underlying I/R injury is still not clear. Recent works showed that circular RNAs play critical roles in many human diseases. In this study, the occurrence of liver I/R injury was validated by an analysis of the blood samples and hematoxylin-eosin (HE) staining of liver tissues. Total RNA was purified and followed by RNA-seq in the purpose of screening the circRNAs in significant differentially expression, which were validated by quantitative PCR. GO and KEGG analysis were performed to determine the function of these differentially expressed circular RNAs. The circular structure of the circRNA was validated with gel electrophoresis and RNase R treatment. We found that some circular RNAs were differentially expressed in Liver I/R mouse models through bioinformatics analysis. These circular RNAs play roles in biological process, cellular component, and molecular function through GO analysis. Meanwhile, Hippo signaling pathway was found to be correlated with circular RNAs function in I/R models by KEGG analysis. To further validate bioinformatics data, two up-regulated and three down-regulated circular RNAs were confirmed in I/R models. The circularity of these differentially expressed circular RNAs was validated through gel electrophoresis and RNase R treatment. In summary, this work provides new insights into the mechanism underlying pathogenesis of liver I/R injury, providing new and potentially efficient targets against I/R injury.
Sigma-Aldrich
AST Activity Assay Kit, sufficient for 100 colorimetric tests
Pricing and availability is not currently available.