ProteoMass™ Insulin chain B oxidized MALDI-MS Standard
Synonym(s):
Insulin Chain B Oxidized from bovine pancreas
Empirical Formula (Hill Notation):
C157H232N40O47S2
CAS Number:
Molecular Weight:
3495.89
Products
Formula Weight
Format
Quality Segment
Package Size
Showing 1-1 of 1
Compare
Product Number
Description
Pricing