
Anti-TJP1 antibody produced in rabbit

Synonym(s):
Anti-Tight junction protein 1 antibody produced in rabbit, Anti-Tight junction protein ZO-1 antibody produced in rabbit, Anti-Zona occludens 1 protein antibody produced in rabbit, Anti-Zonula occludens 1 protein antibody produced in rabbit, Anti-DKFZp686M05161, Anti-MGC133289, Anti-Tight junction protein 1 (zona occludens 1), Anti-ZO-1
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing