
Aervent®-50 Filter Unit 0.2 µm hydrophobic 1/4 in. HB/HB

Synonym(s):
Aervent® 50 Capsule Filter, EPA Est. 041237-FRA-001
Products
Product Category
Brand
Product Line
Length (cm)
Length (in)
Surface Area cm²
Quality Segment
Hydrophobicity
Housing Material
Membrane Material
Pore size (µm)
Sterility
Sterilization Method
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing