
(2-Aminophenyl)acetic acid

Synonym(s):
2-Aminophenylacetic acid
Linear Formula:
H2NC6H4CH2CO2H
CAS Number:
Molecular Weight:
151.16
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing