Merck
  • Indole-Substituted Benzothiazoles and Benzoxazoles as Selective and Reversible MAO-B Inhibitors for Treatment of Parkinson's Disease.
Indole-Substituted Benzothiazoles and Benzoxazoles as Selective and Reversible MAO-B Inhibitors for Treatment of Parkinson's Disease.
ACS chemical neuroscience (2017-03-24)
Min-Ho Nam, Moosung Park, Hyeri Park, Youngjae Kim, Seulki Yoon, Vikram Shahaji Sawant, Ji Won Choi, Jong-Hyun Park, Ki Duk Park, Sun-Joon Min, C Justin Lee, Hyunah Choo
ABSTRACT
To develop novel, selective, and reversible MAO-B inhibitors for safer treatment of Parkinson's disease, benzothiazole and benzoxazole derivatives with indole moiety were designed and synthesized. Most of the synthesized compounds showed inhibitory activities against MAO-B and selectivity over MAO-A. The most active compound was compound 5b, 6-fluoro-2-(1-methyl-1H-indol-5-yl)benzo[d]thiazole with an IC
Sigma-Aldrich
Benzylamine hydrochloride